Spectroquant reference standard for Picco, Move and Multy colorimeters, Supelco®
Lieferant: Merck
1.19301.0001
1.19301.0001EA
150
EUR
1.19301.0001
Spectroquant reference standard for Picco, Move and Multy colorimeters, Supelco®
Standards
Farbreferenzen und Spektroskopie-Standards
A reference standard for chlorine, chlorine dioxide and ozone, that is designed to reassure the user about the accuracy and reliability of the results from the Picco and Multy colorimeters.
Lieferumfang: Standard includes four sealed cells including a Zero cell and stable coloured solutions and is supplied in a handy case with quality data, lot number and expiry date.