Portable colorimeter for disinfection control, Spectroquant® Move Cl₂/O₃/ClO₂/CyA/pH
Lieferant: Merck
1.73635.0001
1.73635.0001EA 668 EUR
1.73635.0001
Portable colorimeter for disinfection control, Spectroquant® Move Cl₂/O₃/ClO₂/CyA/pH
Kolorimeter Mehrparameter-Kolorimeter
The mobile and robust Spectroquant® Move Cl₂/O₃/ClO₂/CyA/pH colorimeter covers five key important parameters for disinfection control (mainly Food and Beverage segment).
  • Simplify your disinfection control
  • Pre-programmed for high quality Spectroquant® test kits
  • Secure rapid reliable results
  • Portable handheld instrument - take the measurement on the spot
Lieferumfang: Instrument supplied in light carrying case with 4× batteries, 3× vials Ø24 mm and 3× vials Ø16 mm, adapter for 16 mm vials, screw driver, guarantee certificate, certificate of compliance, instruction manual.