Reusable earplugs, Rockets®
Lieferant: Moldex
6409
111-0720EA 210 EUR
111-0720
Reusable earplugs, Rockets®
Gehörschutzstöpsel Gehörschutzstöpsel, wiederverwendbar
Corded earplugs made from soft TPE material which is gentle on the skin. The flexible design and the solid handle ensures easy insertion and removal of plugs.
  • Fully detectable
  • Air-cushioned tip
  • Comes with handy PocketPak storage container
Washable and reusable, ideal for the food industry.
SNR: 27 dB
Standards: EN 352-2:2002
Bitte beachten * One size fits all.