Magbeads FastDNA/RNA kit for virus, 192 preparations, MP Biomedicals
Lieferant: MP Biomedicals
MagBeads FastDNA/RNA Kit for Virus is intended for extracting total viral nucleic acid from cell-free/low-cell content biological samples such as body fluids, serum, plasma, soaking solution, tissue homogenate, and culture supernatant.
- High quality viral DNA/ RNA: intact, high yields and free from inhibitors
- High throughput: compatible with a wide range of automated nucleic acid purification platforms
- Flexibility: manual and automation mode of extraction
- Environmentally friendly: no phenol/chloroform or other toxic chemicals.
MagBeads FastDNA/RNA Kit for Virus is based on the purification method of high binding magnetic particles. The sample is lysed and digested with Protease. DNA/RNA is released into the lysate. After addition of magnetic particles and binding solution, DNA/RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.
The adsorbed particles are washed with washing solution to remove proteins and impurities, washed with ethanol to remove salts, and finally DNA/RNA is eluted with nuclease free water.