Adenosine-5ʹ-diphosphate, potassium salt dihydrate ≥98% (durch enzymatische Reinheit), Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
ADP, K
117105-1
117105-1EA
122
EUR
117105-1
Adenosine-5ʹ-diphosphate, potassium salt dihydrate ≥98% (durch enzymatische Reinheit), Millipore®
Adenosine-5ʹ-diphosphate, potassium salt dihydrate
Purity ≥98% by enzymatic assay (dry basis)
Contaminants: AMP: ≤1% and ATP: ≤0.2% by enzymatic assay
Formel:
C₁₀H₁₄N₅O₁₀P₂K·2H₂O Molecular Weight: 501,3 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
72698-48-1 |
Spezifikation Testergebnisse
Form | White solid |
Purity | ≥98% by enzymatic assay (dry basis) |
Contaminants | AMP: ≤1% and ATP: ≤0,2% by enzymatic assay |
Solubility | H2O (50 mg/ml) |
Storage | 2 to 8 °C; Hygroscopic |
Stability | Following reconstitution, aliquot and freeze (−20 °C). Stock solutions are stable for up to 6 months at −20 °C. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...