Drum wrench
Lieferant: JUSTRITE MANUFACTURING
08805
119-0238EA 55.8 EUR
119-0238
Drum wrench
Schraubenschlüssel
Drum Wrench opens drum bung caps.
  • Rugged and non-sparking
  • Design of handle protects the hand
This corrosive-resistant tool fits many 50,8 or 19,05 mm bung styles commonly used on drums.