Porous polymer adsorbent, Tenax®
Lieferant: Merck
11982
11982.EA
453
EUR
11982.
Porous polymer adsorbent, Tenax®
Chromatographie-Sorbentien
GC-Sorbentien
These porous polymers, based on 2,6-diphenyl-p-phenylene oxide, are widely used as an adsorbent in both air collection and purge trap applications.
The unique structure provides alternate and desirable adsorption/desorption characteristics compared to other porous polymers.
Surface area: ~35 m²/g
Density: ~0,25 g/ml (free fall density)