Spill response kit for the laboratory, PIG® HAZ-MAT
Lieferant: NEW PIG
KITE310
121-0568EA 55.75 EUR
121-0568
Spill response kit for the laboratory, PIG® HAZ-MAT
Absorptionsmaterial
Small spill kit suitable chemical spills.
  • Compatible with the most acids and bases including 98% sulphuric acid and 30% sodium hydroxide
  • Sock and mats for containment and absorption
  • Pair of disposable gloves and bags for rapid response and disposal
Lieferumfang: Supplied with 9× absorbent mats (380×510 mm) ; 1× absorbent sock (1,2 m) ; 1× pair 'Silver Shield' chemical gloves ; and 2× plastic disposal bags,1× warning sign and disposal guide.