Baskets for Elmasonic S
Lieferant: ELMA SCHMIDBAUER
1004247
142-0013EA 168 EUR
142-0013
Baskets for Elmasonic S
Körbe Drahtkörbe
These baskets feature a punched pattern on the base and side walls. In conjunction with a set of different pins, these holes allow the objects inside to be positioned as required. For unlimited flexibility. Each item can be individually secured – whether bulky industrial parts, laboratory glassware, small watch components or items of jewellery.
  • Made of stainless steel
  • With Rilsan-coated handles
  • Basket 200 also serves as a holder for sterilisation baskets
The number of objects that can be cleaned in the basket is thus maximised and optimal cleaning results are ensured thanks to perfect placement, which also saves water, cleaning chemicals, energy as well as time and money.