Mini scientific cartridge kit, arium®
Lieferant: Sartorius
H2O-S-PACK
171-0978EA 299 EUR
171-0978
Mini scientific cartridge kit, arium®
Filter Filterkartuschen
Arium® mini scientific pack deionisation cartridge faetures top-down-flow technology which is optimised for the removal of both organic and inorganic constituents. The set was designed specifically to match the unit and delivers ultrapure water that exceeds the ASTM Type 1 quality standard.
  • High performance capacity with efficient ion exchange resins
  • Fast and effective adsorption of impurities through high-grade activated carbon