Cleaning solution for ultrapure water systems, Arium®
Lieferant: Sartorius
611CDS1
171-1024EA 40.1 EUR
171-1024
Cleaning solution for ultrapure water systems, Arium®
Wasseraufbereitungssysteme Reinstwassersysteme
The Arium® cleaning solution is used to effectively remove impurities that can built up over time during water treatment.
  • Highly effective cleaning
  • Free of organic components
  • Gentle to the materials
Regular cleaning not only removes impurities but also leads to a longer service life of consumables and increases the flow rate.
Filled and ready to use in the practical cleaning syringe, the cleaning solution can be used directly.