Diaphragm Pumps, Portable
Lieferant: KNF
85304
181-0074EA 475 EUR
181-0074
Diaphragm Pumps, Portable
Pumpen Vakuum-Membranpumpen
Oil free diaphragm pump designed specifically for use in laboratories and educational establishments. It is an ideal replacement for water jet pumps and is suited for use in applications where vacuum or pressure is required, such as vacuum or pressure filtration. Other applications include gas sampling, circulation and transfer of gases/vapours, flame photometers, vacuum desiccation and small evaporators. The pump is supplied in robust metal cases with a carrying handle and the front panel contains the inlet and outlet connections for 6 mm tubing plus the on/off switch.
  • Designed for continuous operation
  • Maintenance free, quiet running
  • Oil free pumping, so media is not contaminated
  • Environmentally friendly, the pumps use no water so no contaminated waste water is produced