Suction Syphon Pump
Lieferant: NRG DEVELOPMENTS
5915
181-0305EA 34.7 EUR
181-0305
Suction Syphon Pump
Pumpen Fass/Transferpumpen
Suction operated siphon pump.
  • With PE suction tube 390 mm length
  • For drums up to 25 litres