Calcofluor white stain fluoreszierender Farbstoff, für die Mikrobiologie, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
18909-100ML-F
18909-100ML-FEA
66.7
EUR
18909-100ML-F
Calcofluor white stain fluoreszierender Farbstoff, für die Mikrobiologie, Sigma-Aldrich®
Calcofluor white stain
Fluorescent stain for rapid detection of yeasts, fungi and parasitic organisms. Calcofluor white is a non-specific fluorochrom that binds to cellucose and chitin in cell walls.
Composition:
Calcofluor White M2R: 1 g/l
Evans blue: 0,5 g/l