Chromeazurol S (Mordant blue 29), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 2,6-Dichlorodimethyl-sulphoxy-fuchsonedicarboxylic acid sodium salt, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid,trisodium salt, Mordant Blue 29, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid, trisodium salt, Chrome Azurol S

199532-25G
199532-25GEA 116 EUR
199532-25G
Chromeazurol S (Mordant blue 29), Sigma-Aldrich®
Chromeazurol S (Mordant blue 29)

Chromeazurol S has been used to evaluate siderophore production by rhizosphere bacteria.

Formel: C₂₃H₁₆Cl₂O₉S
Molecular Weight: 605,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001615
CAS-Nummer: 1667-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a04dacd4b3ae1c9bc6c8eedcbc99f560674cc99&vid=dffd67acc3a5977f3da1b4eed045d21f5fc26d9f&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR