VWR®, Loading dye base
Lieferant: VWR Chemicals
1B1812-25MLEA 91.5 EUR
1B1812-25ML
VWR®, Loading dye base
Färbemittel Elektrophorese-Färbemittel
This loading dye base is used for custom loading buffer preparation. It is a concentrated solution containing patent blue, bromophenol blue and amaranth tracking dyes, which migrate at approximately 4 000, 400 and 10 bp, on a 1% agarose gel, respectively.
Eliminates the need of weigh messy dye powders.