Capillary tube on foot
Lieferant: GLASWARENFABRIK KARL HECHT
41955010
201-0216EA 41.5 EUR
201-0216
Capillary tube on foot
Lehrmittel Physik-Lehrmittel Mechanics Learning Activities
Five communicating glass tubes with different inner diameters. Used to show capillary forces and how they are affected by the inner diameter of the pipes.
Lieferumfang: Comes with foot.