Distilling bridge with Liebig condenser, Quickfit®
Lieferant: SciLabware
DA1/22
201-0607EA 169 EUR
201-0607
Distilling bridge with Liebig condenser, Quickfit®
Destillationsbauteile Destillierbrücken
PYREX®, borosilicate glass
  • Angle and position pre-defined
  • Less grease contamination
  • No air leakage under vacuum
  • No liquid hold-up at the joints
Comprises Liebig condenser, stillhead and receiver adapter in a single piece.