Slide container
Lieferant: BOETTGER PAUL
08.205.0000
212-0530EA 210 EUR
212-0530
Slide container
Objektträger-Versandbehälter
High-density polyethylene container for microscopy slides.
  • Mailing or storage of microscopy slides
  • For two slides (or four slides stored opposite each other)
  • Shielded ventilation in the cover
Bestellinformation: Containers are supplied without slides.