Carbachol, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: Carbamoylcholinchlorid

212385-100
212385-100EA 98.3 EUR
212385-100
Carbachol, Calbiochem®
Carbachol
Formel: H₂NC(O)O(CH₂)₂N(CH₃)₃Cl
Molecular Weight: 182,65 g/mol
Schmelzpunkt: 207 °C
Dichte: 1,389 g/cm³ (20 °C)
MDL: MFCD00012011
CAS-Nummer: 51-83-2
EINECS: 200-127-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01786

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR