Bariumperchlorat ≥97,0%, AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
21749.236EA
249
EUR
21749.236
Bariumperchlorat ≥97,0%, AnalaR NORMAPUR® analytisches Reagens
Barium perchlorate
Formel:
Ba(ClO₄)₂ Molecular Weight: 336,23 g/mol Schmelzpunkt: 505 °C Dichte: 3,2 g/cm³ (20 °C) Flammpunkt: 21 °C (geschlossener Tiegel) |
MDL:
MFCD00003440 CAS-Nummer: 13465-95-7 EINECS: 236-710-4 UN: 1447 ADR: 5.1 (6.1),II Merck Index: 12,01016 |
Spezifikation Testergebnisse
Assay | Min. 97.0 % |
Heavy metals (as Pb) | Max. 5 ppm |
Insolubility in methanol | Max. 0.1 % |
Not precipitated by H2SO4 (as SO4) | Max. 0.2 % |
Water | Max. 4 % |
Cl (Chloride) | Max. 100 ppm |
Ca (Calcium) | Max. 0.05 % |
Fe (Iron) | Max. 5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...