Ponceau S 0,1 % (w/v) in 5% Essigsäure Färbelösung
Lieferant: Biotium
Synonyme:
3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
22001
22001.EA
96.6
EUR
22001.
Ponceau S 0,1 % (w/v) in 5% Essigsäure Färbelösung
Ponceau S
Ponceau S staining solution is used for the detection of proteins on PVDF and nitrocellulose membranes with a limit of detection of about 250 ng of transferred protein.
- Rapidly stains proteins on membranes pink or light red
- For easy visual inspection of protein transfer
- Reversible and compatible with subsequent Western blotting
Formel:
C₂₂H₁₂N₄Na₄O₁₃S₄ Molecular Weight: 760,56 g/mol |
MDL:
MFCD00003892 CAS-Nummer: 6226-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...