Display trays
Lieferant: CONTACTO
290/290
231-1108EA 39.6 EUR
231-1108
Display trays
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Stainless steel 18/10.
  • With open, flat edge
  • Heavy gauge
  • Mirror polished finish
Multipurpose trays for a wide range of uses. Ideal as display trays.