Blades for safety knives, snap-off, No. 77
Lieferant: MARTOR ARGENTAX
77.60
233-2244EA 12.3 EUR
233-2244
Blades for safety knives, snap-off, No. 77
Rasiermesser und Klingen
Snap-off blades for use with safety cutters.
  • Two-sided ground-edge
  • Double bevel edge-ground
Mountable in:
ARGENTAX JUMBO
ARGENTAX CUTTEX 18 MM
ARGENTAX TAP-O-MATIC