Apiezon® Fett N, Tieftemperatur-Hochvakuumfett halogen- und silikonfrei
Lieferant: M I MATERIALS
FAP/G/NV-00025
24492.136EA 422 EUR
24492.136
Apiezon® Fett N, Tieftemperatur-Hochvakuumfett halogen- und silikonfrei
Schmiermittel
Apiezon N Grease exhibits extremely low vapour pressures at ambient temperatures, 6×10-10 torr at 20 °C, which are further improved by reductions in temperature to the cryogenic region.
  • Can withstand frequent cycling between cryogenic temperatures as low as −273 °C and +30 °C
  • Craze-free at cryogenic temperatures
  • Ability to fill the micropores of adjoining surfaces, so improve thermal contact
  • Powerful 'gettering' action, meaning it absorbs potentially harmful grease and chemical impurities on metal and glass surfaces
  • Easy to clean, use and remove as it is soluble in hydrocarbon and chlorinated solvents
Kryogenes Hoch-Vakuum-Schmierfett. Silikon- und halogenfrei. Ideal für Bürettenhähne und matte Glasoberflächen.