Quarzwolle (~ 9 microns) TECHNICAL
Lieferant: VWR Chemicals
24950.152EA 168 EUR
24950.152
Quarzwolle (~ 9 microns) TECHNICAL
Glas- und Baumwolle
Our silica wool is made from pure fused quartz fiber: amorphous, continuous, tangled, white, odorless, with no volatile component and no shot. Length, shape and arrangement of fibers give the wool its curly appearance preventing compression of the padding and improves its desired insulating properties.
  • High temperature use
  • Resistance to thermal shocks
  • Low thermal conductivity
  • High electrical resistivity
  • Low dielectric constant
  • Good chemical resistance