Natriumpyruvat-Lösung 100 mM with 8.5 g/L NaCl Reagenz für die Zellkultur, Corning®

Lieferant: Corning

Synonyme: 2-Oxopropionsäure Natriumsalz, Brenztraubensäure Natriumsalz, 2-Ketopropionsäure Natriumsalz, α-Ketopropionsäure Natriumsalz

25-000-CI
25-000-CIEA 68.06 EUR
25-000-CI
Natriumpyruvat-Lösung 100 mM with 8.5 g/L NaCl Reagenz für die Zellkultur, Corning®
Natriumpyruvat

100 mM Solution with 8.5 g/L NaCl Sodium pyruvate is often used as a component in culture broth and media as an energy source for propagating cells.


This reagent is also used in Wallen fermentation medium to enhance the conversion of oleic acid to 10-ketostearic acid. It is provided as a 100 mM Solution with 8.5 g/L NaCl.

Formel: CH₃COCOONa
Molecular Weight: 110,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00002586
CAS-Nummer: 113-24-6

Bestellen

Spezifikation Testergebnisse

pH 6.0 ± 1.0
Osmolality 400 ± 100 mOsm/Kg H20
Endotoxin < 0.25 EU/ml@ 1:100
Mycoplasma Tested Negative
Sterility Pass
Cell Growth Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR