illustra™ MicroSpin™ S-400 HR columns
Lieferant: Cytiva
27-5140-01
27-5140-01EA 314 EUR
27-5140-01
illustra™ MicroSpin™ S-400 HR columns
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
illustra™ MicroSpin™ S-400 HR columns designed for rapid purification of PCR products (>200 bp) from unincorporated primers (<32-mers) and nucleotides using spin-column chromatography.
  • Accommodates 25 to 50 μl for post-PCR cleanup prior to cloning or a second amplification reaction or 51 to 100 μl for all other applications
  • Prepacked for convenience with Sephacryl™ S-400 HR pre-equilibriated in TE buffer
  • Ready to use, requiring less than 4 minutes from sample application to collection of purified product
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays