Natriumcarbonat-Decahydrat 99,0-102,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: Soda

27768.298EA 67.4 EUR
27768.298
Natriumcarbonat-Decahydrat 99,0-102,0%, AnalaR NORMAPUR® analytisches Reagens
Natriumcarbonat-Decahydrat
Formel: Na₂CO₃·10H₂O
Molecular Weight: 286,14 g/mol
Schmelzpunkt: 33 °C
Dichte: 1,46 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149178
CAS-Nummer: 6132-02-1
EINECS: 207-838-8

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 102.0 %
Alkali hydroxides and bicarbonates Passes test
Solution in water Passes test
Heavy metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 5 ppm
Total S (as SO4) Max. 30 ppm
Cl (Chloride) Max. 10 ppm
PO4 + SiO4 (as SiO2) Max. 20 ppm
Al (Aluminium) Max. 5 ppm
As (Arsenic) Max. 5 ppm
Ca (Calcium) Max. 20 ppm
Fe (Iron) Max. 2 ppm
K (Potassium) Max. 50 ppm
Mg (Magnesium) Max. 3 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR