1,3-Diiminoisoindoline ≥97%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Isoindoline-1,3-diimine
279528-25G
279528-25GEA
475
EUR
279528-25G
1,3-Diiminoisoindoline ≥97%, Sigma-Aldrich®
1,3-Diiminoisoindoline
1,3-Diiminoisoindoline,97% (1,3-Isoindolinediimine; Phthalimide diimide)
Formel:
C₈H₇N₃ Molecular Weight: 145,17 g/mol Schmelzpunkt: ~197 °C (Zers., Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010062 CAS-Nummer: 57500-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...