Zink, Staub, GPR RECTAPUR® 45 µm

Lieferant: VWR Chemicals
Achtung

29061.290EA 107 EUR
29061.290
Zink, Staub, GPR RECTAPUR® 45 µm
Zinc
Formel: Zn
Molecular Weight: 65,39 g/mol
MDL: MFCD00011291
CAS-Nummer: 7440-66-6
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Assay (total Zn; complexometric) min. 95,0%
Appearance grayish, heavy powder
Substance insoluble in HNO3 max. 0,5%
Cadmium (Cd) max. 0,2%
Copper (Cu) max. 0,005%
Iron (Fe) max. 0,01%
Tin (Sn) max. 0,01%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR