Penicillin : Streptomycin-Lösung 50X, Corning®
Lieferant: Corning
|
Gefahr
|
30-001-CI
30-001-CIEA
117.19
EUR
30-001-CI
Penicillin : Streptomycin-Lösung 50X, Corning®
Penicillin : Streptomycin
This product is a mix of the antibiotics Penicillin (5,000 IU) and Streptomycin (5,000 μg/ mL) in a 50-fold working concentration. Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis. Penicillin-Streptomycin is effective against Gram-negative and Gram-positive bacteria. This antibiotic is recommended for use in cell culture media at 20 ml/L.
Bitte beachten Storage -25°C to -15°C. For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.
CAS-Nummer:
113-98-4 (Penicillin G potassium salt) - 3810-74-0 (Streptomycin sulfate) |
Spezifikation Testergebnisse
Formulation: | |
Penicillin G, potassium salt | 5,000,000 U/L |
Streptomycin sulfate | 5.0 g/l |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...