Dimethylphosphinic chloride ≥97%

Lieferant: Thermo Scientific
Gefahr

30292.02
30292.02EA 111 EUR
30292.02
Dimethylphosphinic chloride ≥97%
Dimethylphosphinic chloride

Purity: 97+% CAS No.: 1111-92-8 Molecular Formula: (CH3)2P(O)Cl Formula Weight: 112.51 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 68° Boiling Point: 204° UN No.: UN3261 MDL No.: MFCD00002078

Formel: C₂H₆ClOP
Molecular Weight: 112,493 g/mol
Siedepunkt: 204 °C
Schmelzpunkt: 68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002078
CAS-Nummer: 1111-92-8
EINECS: 214-187-3
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR