Tetracyclin Hydrochlorid, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

31741-250MG
31741-250MGEA 47.5 EUR
31741-250MG
Tetracyclin Hydrochlorid, Supelco®
Tetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₈
Molecular Weight: 480,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00078142
CAS-Nummer: 64-75-5
EINECS: 200-593-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14daa51192ccc13c7eb90ae6dca126a5feb03381&vid=4172eab3e4a6db07652cf23054be558d1a237c0e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR