Organic reference standard, [D-Asp3, E-Dhb7]-Microcystin-RR solution, analytical standard, Supelco®
Lieferant: Merck
32541-1ML
32541-1MLEA 439 EUR
32541-1ML
Organic reference standard, [D-Asp3, E-Dhb7]-Microcystin-RR solution, analytical standard, Supelco®
Standards Organische Standards
Microcystin is a type of toxin which can adversely affect human health. It is released by cyanobacteria and is commonly found in fresh water sources.
  • [D-Asp3, E-Dhb7]-Microcystin-RR may be used as an analytical standard for the determination of the analyte in:
  • Raw water and drinking water samples by ultra-high pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Water samples, Planktothrix cultures and European crayfish by LC-MS
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=823797940a69f7c8b4b463900a8d5547558bdb9f&vid=823797940a69f7c8b4b463900a8d5547558bdb9f&prid=web&clid=SAPDAM