Fluoresceindiacetat, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Di-O-acetylfluorescein, 3,6-Diacetoxyfluoran, 3-Oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate
343209-5
343209-5EA
114
EUR
343209-5
Fluoresceindiacetat, Sigma-Aldrich®
Fluoresceindiacetat
Formel:
C₂₄H₁₆O₇ Molecular Weight: 416,38 g/mol |
MDL:
MFCD00005062 CAS-Nummer: 596-09-8 EINECS: 209-877-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...