Gentamicinsulfat 50 mg/ml in wässriger Lösung, steril gefiltert, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Gentamicin Sulfatsalz
345815-20ML
345815-20MLEA
207
EUR
345815-20ML
Gentamicinsulfat 50 mg/ml in wässriger Lösung, steril gefiltert, Millipore®
Gentamicinsulfat
Gentamicin sulphate is a broad-spectrum cell culture antibiotic that is non-toxic to viruses and mammalian cells at anti-bacterial and anti-mycoplasmal concentrations. Its extended stability and the slow development of bacterial resistance allow long-term virus and tissue culture studies. Composed of three closely related compounds: gentamycin C1, C1a, and C2.
MDL:
MFCD00270181 CAS-Nummer: 1405-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...