Natriumacetat USP, FCC, J.T.Baker®

Lieferant: Avantor

Synonyme: Essigsäure Natriumsalz

3474-07EA 1020 EUR
3474-07
Natriumacetat USP, FCC, J.T.Baker®
Natriumacetat
Formel: H₃CCOONa
Molecular Weight: 82,03 g/mol
Siedepunkt: 881,4 °C (1013 hPa)
Schmelzpunkt: 324 °C
Dichte: 1,528 g/cm³ (20 °C)
Flammpunkt: >250 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012459
CAS-Nummer: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical 99.0 - 101.0 %
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤0.035 %
USP - Identification Passes Test
USP - Insoluble Matter ≤0.05 %
USP - Loss on Drying at 120°C ≤1.0 %
USP - pH of 3% Solution at 25°C 7.5 - 9.2
USP - Potassium Compounds Passes Test
USP - Residual Acetic Acid, For Information Only For Information Only
USP - Sulfate (SO₄) ≤0.005 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Alkalinity ≤0.2 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying at 120°C ≤1.0 %
FCC - Potassium Compounds Passes Test
Endotoxin Concentration (2.5 EU/g max.) Passes Test
The Class 3 solvent, acetic acid, is used to produce this product.
Residual concentration is controlled to <0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR