VWR® Trichoderma viride Cellulase
Lieferant: VWR Chemicals
390744EEA 456 EUR
390744E
VWR® Trichoderma viride Cellulase
Enzyme
10,000 units is that quantity of enzyme which will degrade 2 cm² of cellulose filter paper (17 mg) in 1 minute in 0,1 M acetate buffer at pH 4,0.