Kulturkolben nach Fernbach, DURAN®
Lieferant: DWK Life Sciences
217746209
391-0310EA 94.4 EUR
391-0310
Kulturkolben nach Fernbach, DURAN®
Kolben Kulturkolben
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Conical shape, straight neck for metal caps
With plain rim.
Typical application: preparation of cultures in nutrient media.