Sample cups for analyser
Lieferant: Greiner Bio-One
668102_1250
391-3720EA 210 EUR
391-3720
Sample cups for analyser
Probenbehälter Probengefäße für Analyser
Cups made of PS.
  • Conical bottom and supported with skirts
  • Good chemical resistance
  • Good temperature resistance
Suitable for analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analysers.