VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Lieferant: VWR Chemicals
392-0442EA
4.9
EUR
392-0442
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Puffer
Gewebekultur-Puffer
Phosphate Buffered Saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate and potassium chloride. PBS helps to maintain a constant pH, is isotonic and non toxic to cells, and provide cells with water and essential inorganic ions. This formulation does not contain calcium and magnesium.
- PBS is used in a variety of laboratory applications:
- Washing cells - prior to DNA/RNA extraction
- Reagent preparation/dilution
- Transporting cells
- Diluent of human biological samples
Note: Please refer to the technical data sheet for recommended use.