ɑ-Cyano-4-hydroxycinnamic acid, ultrarein Matrixsubstanz für MALDI-MS, Supelco®
Lieferant: Merck
|
Achtung
|
Synonyme:
2-Cyano-3-(4-hydroxyphenyl)acrylic Acid, α-Cyano-4-hydroxycinnamic Acid, α-CHCA, CHCA
39468-10X10MG-
39468-10X10MG-EA
329
EUR
39468-10X10MG-
ɑ-Cyano-4-hydroxycinnamic acid, ultrarein Matrixsubstanz für MALDI-MS, Supelco®
alpha-Cyano-4-hydroxycinnamic acid
α-Cyano-4-hydroxycinnamic acid (α-CHCA) functions as matrix and helps in facilitating ionization of proteins and peptides in a matrix-assisted laser desorption/ionization (MALDI) time-of-flight (TOF) mass spectrometer.
Formel:
C₁₀H₇NO₃ Molecular Weight: 189,17 g/mol Schmelzpunkt: 245…250 °C (Lit.) |
MDL:
MFCD00004204 CAS-Nummer: 28166-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...