Schiff-Reagenz, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ500

3952016-500ML
3952016-500MLEA 80.8 EUR
3952016-500ML
Schiff-Reagenz, Sigma-Aldrich®
Schiff-Reagenz

Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections.


Components: Pararosaniline, 1% and sodium metabisulfate, 4%, in hydrochloric acid, 0.25 mol/L


Use Sigma reagent S5133 for staining glycoproteins in polyacrylamide gels.

Dichte: 1,1 g/cm³ (20 °C)
MDL: MFCD00081985
UN: 1789
ADR: 8,III
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR