Ammoniumfluorid 40% in wässriger Lösung
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Fluoroammonium
40292-5L
40292-5LEA
150.2
EUR
40292-5L
Ammoniumfluorid 40% in wässriger Lösung
Ammonium fluoride
Formel:
NH₄F Molecular Weight: 37,04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011423 CAS-Nummer: 12125-01-8 EINECS: 235-185-9 UN: 3287 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...