Periodic table mix 2 for ICP, 17 elements, TraceCERT®, Supelco®
Lieferant: Merck
41135-100ML
41135-100MLEA 275 EUR
41135-100ML
Periodic table mix 2 for ICP, 17 elements, TraceCERT®, Supelco®
Standards Kationischer Standards Mehrelement-Standards
10 mg/L: Au, Ge, Hf, Ir, Mo, Nb, Pd, Pt, Re, Rh, Ru, Sb, Sn, Ta, Ti, W, Zr in 5%hydrochloric acid and 1% hydrofluoric acid (contains HNO₃ traces)
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034