Palladium standard for ICP-MS, TraceCERT®, Supelco®
Lieferant: Merck
42396-100ML
42396-100MLEA
240
EUR
42396-100ML
Palladium standard for ICP-MS, TraceCERT®, Supelco®
Standards
Kationischer Standards
Palladium-Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034