Zink ≥99,8% (durch EDTA Titration), körnig, BAKER ANALYZED® ACS (20 mesh), J.T.Baker®

Lieferant: Avantor
Achtung

4244-07EA 0 EUR
4244-07
Zink ≥99,8% (durch EDTA Titration), körnig, BAKER ANALYZED® ACS (20 mesh), J.T.Baker®
Zinc
Formel: Zn
Molecular Weight: 65,39 g/mol
Siedepunkt: 908 °C (1013 hPa)
Schmelzpunkt: 420 °C
Dichte: 7,14 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011291
CAS-Nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Zn) (by EDTA titrn) ≥99.8 %
Iron (Fe) ≤0.01 %
Lead (Pb) ≤0.01 %
Suitability for Arsenic Determination Passes Test
Trace Impurities - Arsenic (As) ≤0.100 ppm
Mesh - On U.S. No. 16 Sieve ≤5 %
Mesh - Thru U.S. No. 35 Sieve ≤30 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR