Poly(styrene) 2,5% (w/w) Dispersion in Wasser, Latex-Mikrosphären 75.0 micron

Lieferant: Thermo Scientific

42750.AA
42750.AAEA 549 EUR
42750.AA
Poly(styrene) 2,5% (w/w) Dispersion in Wasser, Latex-Mikrosphären 75.0 micron
Poly(styrene)

CAS No.: 9003-53-6 Molecular Formula: [-CH2CH(C6H5)-]n Storage Temperature: -30°C to -10°C Physical Form: Dispersion Appearance: Opalescent Odour: Odourless MDL No.: MFCD00084450

Formel: C₈H₈
Molecular Weight: 125000 - 250000 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00084450
CAS-Nummer: 9003-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR