1,1,1-Triacetoxy-1λ5,2-benziodoxol-3(1H)-on 15% (w/w) in Dichlormethan, AcroSeal®
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester
429001000
429001000.EA
439
EUR
429001000.
1,1,1-Triacetoxy-1λ5,2-benziodoxol-3(1H)-on 15% (w/w) in Dichlormethan, AcroSeal®
Dess-Martin periodinane
Formel:
C₁₃H₁₃IO₈ Molecular Weight: 424,15 g/mol Dichte: 1,362 g/cm³ Lagertemperatur: Kühlschrank |
MDL:
MFCD00130127 CAS-Nummer: 87413-09-0 UN: 1593 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Colorless to light yellow Solution (May develop some turbidity or precipitate) |
Titration Iodometric | 10 to 15 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...